Learn More
Electron Microscopy Sciences Microtome and Cryostat Specimen Disc, 3.8 cm
SDP
Catalog No. 5028801
Encompass_Preferred
Accurately machined chrome plated aluminum discs that fit most microtomes and cryostats. They have a knurled surface for better specimen handling. Straight shank.
Catalog No. 50-288-01 Supplier Electron Microscopy Sciences Supplier No. 6235507
Accurately machined chrome plated aluminum discs that fit most microtomes and cryostats. They have a knurled surface for better specimen handling. Straight shank.