Learn More
Electron Microscopy Sciences Microtome and Cryostat Specimen Disc, 5.1 cm
SDP
Catalog No. 5028804
Encompass_Preferred
Accurately machined chrome plated aluminum discs that fit most microtomes and cryostats. They have a knurled surface for better specimen handling. Straight shank.
Catalog No. 50-288-04 Supplier Electron Microscopy Sciences Supplier No. 6235510
Add to Cart
Add to Cart
Accurately machined chrome plated aluminum discs that fit most microtomes and cryostats. They have a knurled surface for better specimen handling. Straight shank.