Learn More
Leica Biosystems Specimen Disc, Orienting for VT1200 Vibrating Microtome
Catalog No. NC2212240
Encompass
Specimen disc orienting is an accessory for vibrating blade microtomes that enables controlled and reproducible specimen tilting during sectioning. It provides visible angle indicators for precise orientation and is supplied as a single replacement unit.
  • Provides visible 2.5° and 5° angle indicators for reproducible specimen tilting.
  • Enables controlled specimen orientation during sectioning.
  • Supplied as a single unit for easy replacement.
  • Designed for use with vibrating blade microtomes.
This item is not returnable. View return policy
Catalog No. NC2212240 Supplier Leica Biosystems Supplier No. 14048142068
Specimen disc orienting is an accessory for vibrating blade microtomes that enables controlled and reproducible specimen tilting during sectioning. It provides visible angle indicators for precise orientation and is supplied as a single replacement unit.
  • Provides visible 2.5° and 5° angle indicators for reproducible specimen tilting.
  • Enables controlled specimen orientation during sectioning.
  • Supplied as a single unit for easy replacement.
  • Designed for use with vibrating blade microtomes.