Learn More
Leica Biosystems Specimen disc S D 50 mm - specimen holder for Leica microtomes (VT1000 S compatible)
Catalog No. NC0694466
Encompass_Preferred
The Specimen disc S D 50 mm is a 50 mm diameter mounting disc intended to provide a stable surface for biological specimen attachment during sectioning and handling. Check regional availability and ordering information on the manufacturer's product page.
  • Provides a 50 mm diameter mounting surface.
  • Supports stable specimen attachment during sectioning.
  • Sold per unit for flexible ordering.
  • Regional availability may vary; contact supplier for details.
Catalog No. NC0694466 Supplier Leica Biosystems Supplier No. 14046327404
The Specimen disc S D 50 mm is a 50 mm diameter mounting disc intended to provide a stable surface for biological specimen attachment during sectioning and handling. Check regional availability and ordering information on the manufacturer's product page.
  • Provides a 50 mm diameter mounting surface.
  • Supports stable specimen attachment during sectioning.
  • Sold per unit for flexible ordering.
  • Regional availability may vary; contact supplier for details.