Learn More
Leica Biosystems Specimen Disc, 25 mm - For Leica Cryostats (CM1510, CM1800, CM1850, CM1860, CM1900, CM3050)
Catalog No. NC0811724
Encompass
Leica Biosystems specimen disc, 25 mm, is a replacement mounting disc used to secure tissue specimens for cryostat freezing and microtomy. It fits standard specimen chucks and supports stable sample preparation for consistent sectioning.
  • 25 mm diameter for standard specimen mounting.
  • Compatible with common cryostat specimen chucks.
  • Provides a stable mounting surface for consistent sectioning.
  • Single-piece disc designed for reliable specimen adhesion.
  • Intended as a consumable replacement part for routine laboratory use.
Catalog No. NC0811724 Supplier Leica Biosystems Supplier No. 14041619275
Leica Biosystems specimen disc, 25 mm, is a replacement mounting disc used to secure tissue specimens for cryostat freezing and microtomy. It fits standard specimen chucks and supports stable sample preparation for consistent sectioning.
  • 25 mm diameter for standard specimen mounting.
  • Compatible with common cryostat specimen chucks.
  • Provides a stable mounting surface for consistent sectioning.
  • Single-piece disc designed for reliable specimen adhesion.
  • Intended as a consumable replacement part for routine laboratory use.