Learn More
Electron Microscopy Sciences EMS SEM Orbital Specimen Holder Pin Mount
SDP
Catalog No. 501927771
Encompass_Preferred
The EMS Orbital Specimen Holder's clever design allows you to effortlessly rotate the specimen completely around an axis. The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images. The pot is detachable and fully repositionable below the pivoted mount. The position and hold of the sample can be controlled with the simple turn of a screw.
This item is not returnable. View return policy
Catalog No. 50-192-7771 Supplier Electron Microscopy Sciences Supplier No. 75911
The EMS Orbital Specimen Holder's clever design allows you to effortlessly rotate the specimen completely around an axis. The pot is coated to reduce noise, producing a black background behind the subject so every detail stands out, allowing the capture of stunning, clear images. The pot is detachable and fully repositionable below the pivoted mount. The position and hold of the sample can be controlled with the simple turn of a screw.