Learn More
Medicus Health Specimen Transport Insulated Tote, Softside, Shoulder Strap, Medium, Red
SDP
Catalog No. NC3762882
Encompass
• Soft-Sided Specimen Transport Tote with Embroidered Biohazard Symbol, Shoulder Strap, Handle, Medium, Red is an easy and convenient way to transport specimens
• Specimen Cooler is perfect for transporting specimens to and from the lab while maintaining temperatures
• Cooler capacity is 35 quarts
• Heavy-duty, leak-proof 3/4" high density polyurethane foam insulated liner offers superior protection for your specimens or blood products
• Durable leak-resistant insulated liner is easy to clean and disinfect
• Dimensions: 17"W x 10"D x 12"H
• Soft-sided cooler with high-density insulation, a removable shoulder strap, and side pocket
• Twice the insulation of other soft-sided transport coolers
• Made of durable soft canvas material that stands up to travel; features a high-quality leak-proof TPU liner
• Kit this cooler up with our ice pack and blankets to keep your specimens cold (sold separately)
Catalog No. NC3762882 Supplier Medicus Health Supplier No. 2936M2
• Soft-Sided Specimen Transport Tote with Embroidered Biohazard Symbol, Shoulder Strap, Handle, Medium, Red is an easy and convenient way to transport specimens
• Specimen Cooler is perfect for transporting specimens to and from the lab while maintaining temperatures
• Cooler capacity is 35 quarts
• Heavy-duty, leak-proof 3/4" high density polyurethane foam insulated liner offers superior protection for your specimens or blood products
• Durable leak-resistant insulated liner is easy to clean and disinfect
• Dimensions: 17"W x 10"D x 12"H
• Soft-sided cooler with high-density insulation, a removable shoulder strap, and side pocket
• Twice the insulation of other soft-sided transport coolers
• Made of durable soft canvas material that stands up to travel; features a high-quality leak-proof TPU liner
• Kit this cooler up with our ice pack and blankets to keep your specimens cold (sold separately)