Learn More
Electron Microscopy Sciences Duraporter Specimen Transport Box, Red
SDP
Catalog No. 501929439
Encompass_Preferred
The DuraPorter translucent sealed specimen or sample transport tote is an easy-to-load, durable, waterproof, and leak-proof container that securely locks for transport. It is ideal for holding and storing hazardous/sensitive research samples, clinical and phlebotomy samples, blood bags and tissue/organ samples, public health samples, laboratory instruments, and biotransport carrier.
Features
Silicone water-tight seal and three lid clasps to provide secure closure.
Carry handle folds neatly into body space allowing the units to be stacked.
Lid opens a full 180 degree, allowing total access to contents and making it easier to clean.
Accommodates 13 and 16 mm tubes in a 72-place OneRack, or similar sized/brand of rack.
The entire unit can be placed in a freezer or refrigerator.
Polycarbonate construction and is autoclavable.
Color: Red
Catalog No. 50-192-9439 Supplier Electron Microscopy Sciences Supplier No. 62011R
The DuraPorter translucent sealed specimen or sample transport tote is an easy-to-load, durable, waterproof, and leak-proof container that securely locks for transport. It is ideal for holding and storing hazardous/sensitive research samples, clinical and phlebotomy samples, blood bags and tissue/organ samples, public health samples, laboratory instruments, and biotransport carrier.
Features
Silicone water-tight seal and three lid clasps to provide secure closure.
Carry handle folds neatly into body space allowing the units to be stacked.
Lid opens a full 180 degree, allowing total access to contents and making it easier to clean.
Accommodates 13 and 16 mm tubes in a 72-place OneRack, or similar sized/brand of rack.
The entire unit can be placed in a freezer or refrigerator.
Polycarbonate construction and is autoclavable.
Color: Red