Learn More
Medicus Health Specimen Transport Cooler, 12V Plug-In, 18 Quart
SDP
Catalog No. NC3806000
Encompass
• 18 Quart Thermoelectric Cooler-12 V- provides an "active" cooling solution for the transport of laboratory specimens
• Slim design fits well in cars, especially behind seats or between front and back seats, making it convenient for travel
• Each unit will cool to approximately 40° F lower than ambient temperature and can also heat up to approximately 135° F
• Comes complete with 12-volt power plug for vehicle
• Overall dimensions: 16"L x 8.5"W x 17.25"H
• 18 Quart lightweight coolers have convenient transport handle
• NOTE: Cooler does not lock
Catalog No. NC3806000 Supplier Medicus Health Supplier No. 2734M1
• 18 Quart Thermoelectric Cooler-12 V- provides an "active" cooling solution for the transport of laboratory specimens
• Slim design fits well in cars, especially behind seats or between front and back seats, making it convenient for travel
• Each unit will cool to approximately 40° F lower than ambient temperature and can also heat up to approximately 135° F
• Comes complete with 12-volt power plug for vehicle
• Overall dimensions: 16"L x 8.5"W x 17.25"H
• 18 Quart lightweight coolers have convenient transport handle
• NOTE: Cooler does not lock