Learn More
Medicus Health Specimen Transport Tote, Softside, With Shoulder Strap, Medium, Blue
SDP
Catalog No. NC1802924
Encompass
∙ Soft-Sided Insulated Specimen Transport Tote with Biohazard Label, Shoulder Strap and Handle Medium Size is an easy and convenient way to transport specimens ∙Specimen Cooler is perfect for transporting specimens to and from the lab ∙Tote comes with premium raised vinyl BioHazard Label which helps clearly and professionally labels tote contents ∙Heavy-duty, leak-proof 3/4" high density polyurethane foam insulated liner that's easy to clean & offers superior protection for your specimens or blood products∙External Dimensions: 17"W x 10"D x 12"H∙Soft-sided cooler with high-density insulation, a removable shoulder strap, and side pocket∙Twice the insulation of other soft-sided transport coolers∙Features a high-quality leak-proof TPU liner∙Convenient transport handle and padded should strap with heavy-duty zipper to secure contents
This item is not returnable. View return policy
Catalog No. NC1802924 Supplier Medicus Health Supplier No. 2936M2BLUE
∙ Soft-Sided Insulated Specimen Transport Tote with Biohazard Label, Shoulder Strap and Handle Medium Size is an easy and convenient way to transport specimens ∙Specimen Cooler is perfect for transporting specimens to and from the lab ∙Tote comes with premium raised vinyl BioHazard Label which helps clearly and professionally labels tote contents ∙Heavy-duty, leak-proof 3/4" high density polyurethane foam insulated liner that's easy to clean & offers superior protection for your specimens or blood products∙External Dimensions: 17"W x 10"D x 12"H∙Soft-sided cooler with high-density insulation, a removable shoulder strap, and side pocket∙Twice the insulation of other soft-sided transport coolers∙Features a high-quality leak-proof TPU liner∙Convenient transport handle and padded should strap with heavy-duty zipper to secure contents