Learn More
Alkali Scientific ASI™ Specimen Vial – 7mL, PP
SDP
Catalog No. 503028254
Encompass_Preferred
7mL Specimen Vial are supplied in translucent, virgin polypropylene or crystal clear virgin polystyrene. Both versions are supplied with an attached polyethylene screw cap. They are CE marked and certified leak proof. Packaged 700 vials with attached caps per case. Virgin Polypropylene/Polystyrene Non-Cytotoxic Leakproof Cap Economical Bulk Packaging
Catalog No. 50-302-8254 Supplier Alkali Scientific Supplier No. SV251
7mL Specimen Vial are supplied in translucent, virgin polypropylene or crystal clear virgin polystyrene. Both versions are supplied with an attached polyethylene screw cap. They are CE marked and certified leak proof. Packaged 700 vials with attached caps per case. Virgin Polypropylene/Polystyrene Non-Cytotoxic Leakproof Cap Economical Bulk Packaging