Learn More
Biotage SPECLMN C18EC 10G/70ML 16
SDP
Catalog No. 502311658
Encompass
Column, SPE; Biotage; Isolute; C18; End Capped; 10g/70mL; 16/pk.
Catalog No. 50-231-1658 Supplier Biotage Supplier No. 2211000F
Column, SPE; Biotage; Isolute; C18; End Capped; 10g/70mL; 16/pk.