Learn More
Biotage SPECLMN DFR 20U/3ML 100
SDP
Catalog No. 502311345
Encompass
Column, Filtration; Biotage; Isolute: For SPE; Double fritted; Analytical grade; 20um frits; 3mL; 100/pack
Catalog No. 50-231-1345 Supplier Biotage Supplier No. 1201022B
Column, Filtration; Biotage; Isolute: For SPE; Double fritted; Analytical grade; 20um frits; 3mL; 100/pack