Learn More
Biotage SPECLMN ENV+ 10MG/1ML 100
SDP
Catalog No. 502312381
Encompass
Column, SPE; Biotage; Isolute; ENV+; Standard; Reservoir volume: 1mL; Media weight: 10mg; 100/pack
Catalog No. 50-231-2381 Supplier Biotage Supplier No. 9150001A
Column, SPE; Biotage; Isolute; ENV+; Standard; Reservoir volume: 1mL; Media weight: 10mg; 100/pack