Learn More
Biotage SPECLMN ENV+ 200MG/6ML 30
SDP
Catalog No. 502312401
Encompass
Column, SPE; Biotage; Isolute; ENV+; Reservoir volume: 6mL; Media weight: 200mg; 30/pack
Catalog No. 50-231-2401 Supplier Biotage Supplier No. 9150020C
Column, SPE; Biotage; Isolute; ENV+; Reservoir volume: 6mL; Media weight: 200mg; 30/pack