Learn More
Biotage SPECLMN ENV+ 500MG/3ML 50
SDP
Catalog No. 502312408
Encompass
Column, SPE; Biotage; Isolute; ENV+; Reservoir volume: 3mL; Media weight: 500mg; 50/pack
Catalog No. 50-231-2408 Supplier Biotage Supplier No. 9150050B
Column, SPE; Biotage; Isolute; ENV+; Reservoir volume: 3mL; Media weight: 500mg; 50/pack