Learn More
Biotage SPECLMN FL 100MG/1ML 100
SDP
Catalog No. 502312205
Encompass
Column, SPE; Biotage; Isolute; Florisil; Reservoir volume: 1mL; Media weight: 100mg; 100/pack
Catalog No. 50-231-2205 Supplier Biotage Supplier No. 7120010A
Column, SPE; Biotage; Isolute; Florisil; Reservoir volume: 1mL; Media weight: 100mg; 100/pack