Learn More
Biotage SPECLMN NH2 100MG/1ML 100
SDP
Catalog No. 502311947
Encompass
Column, SPE; Biotage; Isolute; NH2; Reservoir volume: 1mL; Media weight: 100mg; 100/pack
Catalog No. 50-231-1947 Supplier Biotage Supplier No. 4700010A
Column, SPE; Biotage; Isolute; NH2; Reservoir volume: 1mL; Media weight: 100mg; 100/pack