Learn More
Biotage SPECLMN NH2 200MG/3ML 50
SDP
Catalog No. 502311951
Encompass
Column, SPE; Biotage; Isolute; NH2; Reservoir volume: 3mL; Media weight: 200mg; 50/pack
Catalog No. 50-231-1951 Supplier Biotage Supplier No. 4700020B
Column, SPE; Biotage; Isolute; NH2; Reservoir volume: 3mL; Media weight: 200mg; 50/pack