Learn More
Biotage SPECLMN PEAX 500MG/6ML 30
SDP
Catalog No. 502312036
Encompass
Column, SPE; Biotage; Isolute; PE-AX; Reservoir volume: 6mL; Media weight: 500mg; 30/pack
Catalog No. 50-231-2036 Supplier Biotage Supplier No. 5030050C
Column, SPE; Biotage; Isolute; PE-AX; Reservoir volume: 6mL; Media weight: 500mg; 30/pack