Learn More
Biotage SPECLMN SCX 50MG/1ML 100
SDP
Catalog No. 502312084
Encompass
Column, SPE; Biotage; Isolute; SCX; Reservoir volume: 1mL; Media weight: 50mg; 100/pack
Catalog No. 50-231-2084 Supplier Biotage Supplier No. 5300005A
Column, SPE; Biotage; Isolute; SCX; Reservoir volume: 1mL; Media weight: 50mg; 100/pack