Learn More
Biotage SPECLMN SCX2 5G/25ML 20
SDP
Catalog No. 502312139
Encompass
Column, SPE; Biotage; Isolute; SCX-2; Reservoir volume: 25mL; Media weight: 2g; 20/pack
Catalog No. 50-231-2139 Supplier Biotage Supplier No. 5320500E
Column, SPE; Biotage; Isolute; SCX-2; Reservoir volume: 25mL; Media weight: 2g; 20/pack