Learn More
Biotage SPECLMN SFRIT 20U/10ML 50
SDP
Catalog No. 502311411
Encompass
Reservoir, Single fritted; Biotage; Isolute; SPE; Analytical grade; XL (G); Filtration columns; PE; Volume: 10mL; 20um, Frits, G; 50/pk.
Catalog No. 50-231-1411 Supplier Biotage Supplier No. 1201124G
Reservoir, Single fritted; Biotage; Isolute; SPE; Analytical grade; XL (G); Filtration columns; PE; Volume: 10mL; 20um, Frits, G; 50/pk.