Learn More
Biotage SPECLMN SFRIT 20U/70ML 50
SDP
Catalog No. 502311415
Encompass
Reservoir, Single fritted; Biotage; Isolute; SPE; Analytical grade; Filtration columns; PE; Volume: 70mL; 20um, Frits, D; 50/pk.
Catalog No. 50-231-1415 Supplier Biotage Supplier No. 1201128F
Reservoir, Single fritted; Biotage; Isolute; SPE; Analytical grade; Filtration columns; PE; Volume: 70mL; 20um, Frits, D; 50/pk.