Learn More
Biotage SPECLMN SILICA 100MG/1ML 100
SDP
Catalog No. 502311919
Encompass
Column, SPE; Biotage; Isolute; Si; Reservoir volume: 1mL; Media weight: 100mg; 100/pack
Catalog No. 50-231-1919 Supplier Biotage Supplier No. 4600010A
Column, SPE; Biotage; Isolute; Si; Reservoir volume: 1mL; Media weight: 100mg; 100/pack