Learn More
Biotage SPECLMNDF DFR 20U/15ML 100
SDP
Catalog No. 502311350
Encompass
Column, Filtration; Biotage; Isolute: For SPE; Double fritted; Analytical grade; 20um frits; 15mL; 100/pack
Catalog No. 50-231-1350 Supplier Biotage Supplier No. 1201025D
Column, Filtration; Biotage; Isolute: For SPE; Double fritted; Analytical grade; 20um frits; 15mL; 100/pack