Learn More
Biotage SPECLMNL FL 500MG/6ML 30
SDP
Catalog No. 502312216
Encompass
Column, SPE; Biotage; Isolute; Florisil; Glass; Reservoir volume: 6mL; Media weight: 500mg; 30/pack
Catalog No. 50-231-2216 Supplier Biotage Supplier No. 7120050L
Column, SPE; Biotage; Isolute; Florisil; Glass; Reservoir volume: 6mL; Media weight: 500mg; 30/pack