Learn More
Biotage SPECLMNSTD DFR 20U/70ML 50
SDP
Catalog No. 502311443
Encompass
Column, Filtration; Biotage; SPE; Double fritted; Standard grade; 2 x 20um Frits, F; Capacity: 70mL; 50/pk.
Catalog No. 50-231-1443 Supplier Biotage Supplier No. 1201217F
Column, Filtration; Biotage; SPE; Double fritted; Standard grade; 2 x 20um Frits, F; Capacity: 70mL; 50/pk.