Learn More
Biotage SPECLMNTL 20U/6ML 100
SDP
Catalog No. 502311398
Encompass
Reservoir, Empty; Biotage; Isolute; Array-24 system; For SPE columns; Single fritted; Standard grade; Tabless; Polyethylene; 6mL; 20um; 100/pack
Catalog No. 50-231-1398 Supplier Biotage Supplier No. 1201113CG
Reservoir, Empty; Biotage; Isolute; Array-24 system; For SPE columns; Single fritted; Standard grade; Tabless; Polyethylene; 6mL; 20um; 100/pack