Learn More
Biotage SPECLMNXL NH2 500MG/10ML 50
SDP
Catalog No. 502311958
Encompass
Column, SPE; Biotage; Isolute; NH2; Reservoir volume: 10mL; Media weight: 500mg; 30/pack
Catalog No. 50-231-1958 Supplier Biotage Supplier No. 4700050H
Column, SPE; Biotage; Isolute; NH2; Reservoir volume: 10mL; Media weight: 500mg; 30/pack