Learn More
Biotage SPECLNM MULTIMODE 500MG/3ML 50
SDP
Catalog No. 502312346
Encompass
Column, SPE; Biotage; Isolute; Multimode; Reservoir volume: 3mL; Media weight: 500mg; 50/pack
Catalog No. 50-231-2346 Supplier Biotage Supplier No. 9040050B
Column, SPE; Biotage; Isolute; Multimode; Reservoir volume: 3mL; Media weight: 500mg; 50/pack