Learn More
Chemglass Life Sciences SpecMate Vis Spectrophotometer
SDP
Catalog No. 501311338
Encompass
SpecMate Vis Spectrophotometer. • Large LCD Screen• Auto Zero and Blank• USB & Parallel Printer Port• Pre-Aligned Design for Easy Lamp Change• Optional Test Tube Holder, 8-22mm O.D.• Sample Compartment Holds 4-10mm Square Cuvettes• Software Included Allows for Kinetics and Protein WorkPrecise design spectrophotometer with manual wavelength setting and a large LCD display. This high quality instrument ensures excellent performance and is widely used in educational and analysis laboratories. Wavelength can be read directly from the screen.Unit is supplied with 4 glass 10mm square Cuvettes and basic software that is used for quantitative analysis and kinetics.
Catalog No. 50-131-1338 Supplier Chemglass Life Sciences Supplier No. CLS404401
SpecMate Vis Spectrophotometer. • Large LCD Screen• Auto Zero and Blank• USB & Parallel Printer Port• Pre-Aligned Design for Easy Lamp Change• Optional Test Tube Holder, 8-22mm O.D.• Sample Compartment Holds 4-10mm Square Cuvettes• Software Included Allows for Kinetics and Protein WorkPrecise design spectrophotometer with manual wavelength setting and a large LCD display. This high quality instrument ensures excellent performance and is widely used in educational and analysis laboratories. Wavelength can be read directly from the screen.Unit is supplied with 4 glass 10mm square Cuvettes and basic software that is used for quantitative analysis and kinetics.