Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Absorbance and Fluorescence, Top/Bottom, 96/384
Catalog No. 5000014377
Encompass_Preferred
The SpectraMax M2e is a configurable multi-mode microplate reader designed for absorbance, fluorescence, time-resolved fluorescence, and luminescence assays. It includes SoftMax Pro software for data acquisition and analysis, supports top and bottom reads, and is compatible with a wide range of microplate formats and automation workflows.
  • Configurable for absorbance, fluorescence intensity, time-resolved fluorescence, and luminescence detection.
  • Supports 6, 12, 24, 48, 96, and 384 well microplates.
  • Dual monochromators tunable in 1 nm increments for flexible wavelength selection.
  • Wavelength coverage suitable for UV-visible and fluorescence assays (absorbance 200-1000 nm, fluorescence 250-850 nm).
  • Includes temperature control to ambient +4°C to 45°C, linear shaking, and automation compatibility.
  • Compact footprint and laboratory weight suitable for benchtop placement (≈22 x 58 x 38 cm, ≈35 lb).
Catalog No. 50-000-14377 Supplier Molecular Devices Supplier No. M2E
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M2e is a configurable multi-mode microplate reader designed for absorbance, fluorescence, time-resolved fluorescence, and luminescence assays. It includes SoftMax Pro software for data acquisition and analysis, supports top and bottom reads, and is compatible with a wide range of microplate formats and automation workflows.
  • Configurable for absorbance, fluorescence intensity, time-resolved fluorescence, and luminescence detection.
  • Supports 6, 12, 24, 48, 96, and 384 well microplates.
  • Dual monochromators tunable in 1 nm increments for flexible wavelength selection.
  • Wavelength coverage suitable for UV-visible and fluorescence assays (absorbance 200-1000 nm, fluorescence 250-850 nm).
  • Includes temperature control to ambient +4°C to 45°C, linear shaking, and automation compatibility.
  • Compact footprint and laboratory weight suitable for benchtop placement (≈22 x 58 x 38 cm, ≈35 lb).