Learn More
Molecular Devices Molecular Devices Microplate Reader, Dual Monochromator, 96/384-Well, Dual-Mode Cuvette
Catalog No. 5000014684
Encompass_Preferred
The SpectraMax M2e multi-mode microplate reader provides absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF measurements with flexible plate and cuvette workflows. It features dual monochromators and tunable wavelength selection for assay development and routine screening.
  • Measures absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF.
  • Compatible with 6-384 well microplates for assay format flexibility.
  • Dual monochromators with 1 nm wavelength increments for precise wavelength selection.
  • Wavelength coverage for absorbance 200-1000 nm, fluorescence excitation 250-850 nm, and emission 250-850 nm.
  • Includes a dual-mode cuvette port for top and bottom read formats.
  • Operates from 100-240 Vac, 50/60 Hz, 3 A power input.
  • Designed for integration with automation and robotic systems.
Catalog No. 50-000-14684 Supplier Molecular Devices Supplier No. M2EG
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M2e multi-mode microplate reader provides absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF measurements with flexible plate and cuvette workflows. It features dual monochromators and tunable wavelength selection for assay development and routine screening.
  • Measures absorbance, fluorescence, luminescence, time-resolved fluorescence, fluorescence polarization, and HTRF.
  • Compatible with 6-384 well microplates for assay format flexibility.
  • Dual monochromators with 1 nm wavelength increments for precise wavelength selection.
  • Wavelength coverage for absorbance 200-1000 nm, fluorescence excitation 250-850 nm, and emission 250-850 nm.
  • Includes a dual-mode cuvette port for top and bottom read formats.
  • Operates from 100-240 Vac, 50/60 Hz, 3 A power input.
  • Designed for integration with automation and robotic systems.