Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Monochromator, 6-384 Well, 200-1000 nm
Catalog No. 5000014686
Encompass_Preferred
The SpectraMax M3 is a multi-mode microplate reader for absorbance, fluorescence, and luminescence assays. It combines dual monochromator optics, a dedicated luminescence PMT, integrated temperature control, and is operated with preloaded data acquisition and analysis software.
  • Measures absorbance (200-1000 nm), fluorescence, and luminescence (250-850 nm).
  • Compatible with 6, 12, 24, 48, 96, and 384-well microplates.
  • Dual monochromators with 1 nm wavelength increments for precise selection.
  • Dedicated PMT for low-noise luminescence detection and high fluorescence sensitivity.
  • Integrated temperature control from ambient +2°C to 60°C with ±1°C accuracy.
  • Xenon flash lamp light source and programmable orbital shaker for assay mixing.
  • Robotics compatible design for automation and high-throughput workflows.
Catalog No. 50-000-14686 Supplier Molecular Devices Supplier No. M3G
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M3 is a multi-mode microplate reader for absorbance, fluorescence, and luminescence assays. It combines dual monochromator optics, a dedicated luminescence PMT, integrated temperature control, and is operated with preloaded data acquisition and analysis software.
  • Measures absorbance (200-1000 nm), fluorescence, and luminescence (250-850 nm).
  • Compatible with 6, 12, 24, 48, 96, and 384-well microplates.
  • Dual monochromators with 1 nm wavelength increments for precise selection.
  • Dedicated PMT for low-noise luminescence detection and high fluorescence sensitivity.
  • Integrated temperature control from ambient +2°C to 60°C with ±1°C accuracy.
  • Xenon flash lamp light source and programmable orbital shaker for assay mixing.
  • Robotics compatible design for automation and high-throughput workflows.