Learn More
Medchemexpress LLC HY-N6075 5mg Medchemexpress, Specneuzhenide CAS:449733-84-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60755MG
Encompass_Preferred
Medchemexpress, HY-N6075 5mg Specneuzhenide CAS:449733-84-0 Specneuzhenide (Nuezhenide) is a phenol glycoside isolated from Ligustrum sinense. Specneuzhenide (Nuezhenide) possesses anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0448