Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Specnuezhenide ((8E)-Nuezhenide) | 39011-92-2 | 686.66 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN06655MG

Encompass_Preferred

Specnuezhenide ((8E)-Nuezhenide) is a compound isolated from the fruits of *Ligustrum lucidum*. It demonstrates the ability to inhibit IL-1β-induced inflammation in chondrocytes by suppressing NF-κB and wnt/β-catenin signaling pathways. Additionally, Specnuezhenide exhibits anti-inflammatory effects in a rat model of osteoarthritis (OA).

  • Isolated from *Ligustrum lucidum* fruits.
  • Inhibits IL-1β-induced inflammation in chondrocytes.
  • Exerts anti-inflammatory effects in osteoarthritis rat models.
  • For research use only.
  • Purity of 99.52%.
  • Appears as a solid.
  • Color is white to off-white.

Catalog No. 50-003-88816


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.