Learn More
Medchemexpress LLC Specnuezhenide ((8E)-Nuezhenide) | 39011-92-2 | 99.5% | 686.66 g/mol | C31H42O17 | 10 MG
SDP
Catalog No. 5000296069
Encompass_Preferred
Specnuezhenide is a natural iridoid glycoside isolated from Ligustrum lucidum. It exhibits activity against NF-κB and Wnt/β-catenin signaling and is provided as a high-purity research reagent for in-vitro and analytical applications.
  • Natural iridoid glycoside
  • Modulates NF-κB and Wnt/β-catenin signaling
  • High purity (~99.5%), suitable for analytical and in vitro studies
  • Available in small research pack sizes
  • Appropriate for biochemical and cell-based assays
Catalog No. 50-002-96069 Supplier Medchemexpress LLC Supplier No. HYN066510MG
Specnuezhenide is a natural iridoid glycoside isolated from Ligustrum lucidum. It exhibits activity against NF-κB and Wnt/β-catenin signaling and is provided as a high-purity research reagent for in-vitro and analytical applications.
  • Natural iridoid glycoside
  • Modulates NF-κB and Wnt/β-catenin signaling
  • High purity (~99.5%), suitable for analytical and in vitro studies
  • Available in small research pack sizes
  • Appropriate for biochemical and cell-based assays