Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Spectacle Storage Cases

Catalog No. 19151627 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Product Type:
Eyewear Case
Color:
Black
Blue
For Use With (Application):
For storing glasses
Ideal for lens cleaning
Description:
Drawstring Storage Case
Zippered Storage Case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color For Use With (Application) Description
19-151-627 Eyewear Case Blue Ideal for lens cleaning Zippered Storage Case
19-151-628 Eyewear Case Black For storing glasses Drawstring Storage Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-151-627 Supplier Pyramex™ Supplier No. CA200B
Add to Cart
Add to Cart

Microfiber cases for glasses ideal for lens cleaning

  • Available in black
  • Available with zipper or drawstring

Specifications

Description Zippered Storage Case
Color Blue
Product Type Eyewear Case
For Use With (Equipment) For glasses
For Use With (Application) Ideal for lens cleaning
Material Microfiber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.