Learn More
Apexbio Technology LLC Spectinomycin dihydrochloride 21736-83-4 5g
SDP
Catalog No. 5000063752
Encompass_Preferred
Spectinomycin dihydrochloride is an aminocyclitol antibiotic derived from Streptomyces spectabilis exerting antibacterial activity by selectively binding to the bacterial 30S ribosomal subunit and disrupting protein synthesis initiation and elongation steps Due to its defined mechanism impacting gram-negative and certain gram-positive bacteria spectinomycin dihydrochloride is commonly utilized in microbiology and cell biology studies for selective inhibition assays bacterial resistance investigations and functional analysis of ribosomal function Experimental studies typically report inhibitory effects against sensitive bacterial strains with IC50 values ranging approximately from 5 to 20 g/mL depending on bacterial species and experimental methodologies It is suitable for research applications involving bacterial cultures protein translation assays and antibiotic resistance profiling
Catalog No. 50-000-63752 Supplier Apexbio Technology LLC Supplier No. B33425000
Spectinomycin dihydrochloride is an aminocyclitol antibiotic derived from Streptomyces spectabilis exerting antibacterial activity by selectively binding to the bacterial 30S ribosomal subunit and disrupting protein synthesis initiation and elongation steps Due to its defined mechanism impacting gram-negative and certain gram-positive bacteria spectinomycin dihydrochloride is commonly utilized in microbiology and cell biology studies for selective inhibition assays bacterial resistance investigations and functional analysis of ribosomal function Experimental studies typically report inhibitory effects against sensitive bacterial strains with IC50 values ranging approximately from 5 to 20 g/mL depending on bacterial species and experimental methodologies It is suitable for research applications involving bacterial cultures protein translation assays and antibiotic resistance profiling