Learn More
Crescent Chemical Co Inc SPECTINOMYCIN-2HCL PENTAHYDRAT
SDP
Catalog No. 50248154
Encompass_Preferred
Spectinomycin-2HCl pentahydrate research grade Cell culture tested.Water-soluble aminoglycoside antibiotic from Streptomyces sp. (1,2). Inhibitor of protein synthesis (3). Differs from other aminoglycoside derivatives in that it is bacteriostatic instead of bactericidal.Stock so
Catalog No. 50-248-154 Supplier Crescent Chemical Co Inc Supplier No. SE35294.01
Spectinomycin-2HCl pentahydrate research grade Cell culture tested.Water-soluble aminoglycoside antibiotic from Streptomyces sp. (1,2). Inhibitor of protein synthesis (3). Differs from other aminoglycoside derivatives in that it is bacteriostatic instead of bactericidal.Stock so