Learn More
AG Scientific Inc Spectinomycin 5 G
Supplier:  AG Scientific Inc S28735GM
Encompass_Preferred
Spectinomycin dihydrochloride pentahydrate is an aminocyclitol antibiotic that inhibits protein synthesis. Cell culture tested.
Catalog No. 50-226-8389