Learn More
Medchemexpress LLC HY-B1828A 500mg Medchemexpress, Spectinomycin (dihydrochloride pentahydrate) CAS:22189-32-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1828A500MG
Encompass_Preferred
Medchemexpress, HY-B1828A 500mg Spectinomycin (dihydrochloride pentahydrate) CAS:22189-32-8 Spectinomycin dihydrochloride pentahydrate is a broad-spectrum aminocyclitol antibiotic that inhibits the growth of a variety of gram-positive and gram-negative organisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2713