Learn More
Gold Biotechnology Inc Spectinomycin Dihydrochloride Pentahydrate, USP Grade 50g
SDP
Supplier:  Gold Biotechnology Inc S1405050G
Encompass_Preferred
Spectinomycin is an aminocyclitol antibiotic isolated from Streptomyces spectabilis. It is effective against gram-positive and gram-negative bacteria, but most commonly used against Gonococcus species. It has also been used to monitor cell fate during leaf development and as a selection marker in plants to display successful transformation of the Spcr gene.
Catalog No. NC1506839