Learn More
Gold Biotechnology Inc Spectinomycin Dihydrochloride Pentahydrate, USP Grade 50 g
SDP
Catalog No. 501532835
Encompass_Preferred
Spectinomycin is an aminocyclitol antibiotic isolated from Streptomyces spectabilis. It is effective against gram-positive and gram-negative bacteria, but most commonly used against Gonococcus species. It has also been used to monitor cell fate during leaf development and as a selection marker in plants to display successful transformation of the Spcr gene.
Catalog No. 50-153-2835 Supplier Gold Biotechnology Inc Supplier No. S14050
Spectinomycin is an aminocyclitol antibiotic isolated from Streptomyces spectabilis. It is effective against gram-positive and gram-negative bacteria, but most commonly used against Gonococcus species. It has also been used to monitor cell fate during leaf development and as a selection marker in plants to display successful transformation of the Spcr gene.