Learn More
Medchemexpress LLC Spectinomycin dihydrochloride | 21736-83-4 | 99.7% | 1 ML
SDP
Catalog No. 5000408078
Encompass_Preferred
Spectinomycin dihydrochloride is a broad-spectrum antibiotic that inhibits the growth of various gram-positive and gram-negative organisms. It acts by selectively targeting the bacterial ribosome, thereby interrupting protein synthesis. It is also a noncompetitive inhibitor of td intron RNA with a Ki value of 7.2 mM.
  • Inhibits protein synthesis in cells and Escherichia coli extracts
  • Blocks the translocation of peptidyl-tRNAs from the A-site to the P-site
  • Exhibits splicing inhibition dependent on pH changes and Mg2+ concentration
Catalog No. 50-004-08078 Supplier Medchemexpress LLC Supplier No. HYB043810MM/1ML
Spectinomycin dihydrochloride is a broad-spectrum antibiotic that inhibits the growth of various gram-positive and gram-negative organisms. It acts by selectively targeting the bacterial ribosome, thereby interrupting protein synthesis. It is also a noncompetitive inhibitor of td intron RNA with a Ki value of 7.2 mM.
  • Inhibits protein synthesis in cells and Escherichia coli extracts
  • Blocks the translocation of peptidyl-tRNAs from the A-site to the P-site
  • Exhibits splicing inhibition dependent on pH changes and Mg2+ concentration