Learn More
Medchemexpress LLC Spectinomycin dihydrochloride | 21736-83-4 | 99.73% | 25 G
SDP
Catalog No. 5000407375
Encompass_Preferred
Spectinomycin dihydrochloride is a broad-spectrum antibiotic that inhibits the growth of various gram-positive and gram-negative organisms. It functions by selectively targeting bacterial ribosomes and interrupting protein synthesis, and acts as a noncompetitive inhibitor of td intron RNA with a Ki value of 7.2 mM.
  • Broad-spectrum antibiotic
  • Inhibits growth of gram-positive and gram-negative organisms
  • Selectively targets bacterial ribosomes
  • Interrupts protein synthesis
  • Acts as a noncompetitive inhibitor of td intron RNA
  • Inhibits polypeptide synthesis
  • Blocks translocation of peptidyl-tRNAs
Catalog No. 50-004-07375 Supplier Medchemexpress LLC Supplier No. 25GHYB0438
Spectinomycin dihydrochloride is a broad-spectrum antibiotic that inhibits the growth of various gram-positive and gram-negative organisms. It functions by selectively targeting bacterial ribosomes and interrupting protein synthesis, and acts as a noncompetitive inhibitor of td intron RNA with a Ki value of 7.2 mM.
  • Broad-spectrum antibiotic
  • Inhibits growth of gram-positive and gram-negative organisms
  • Selectively targets bacterial ribosomes
  • Interrupts protein synthesis
  • Acts as a noncompetitive inhibitor of td intron RNA
  • Inhibits polypeptide synthesis
  • Blocks translocation of peptidyl-tRNAs