Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectinomycin Dihydrochloride, Fisher BioReagents™

Catalog No. BP29571 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP29571 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP29571

missing translation for 'validMainframeMsg'

Inhibits protein synthesis | CAS: 22189-32-8 | C14H36Cl2N2O12 | 495.34 g/mol

Inhibits protein synthesis

TRUSTED_SUSTAINABILITY

Specifications

Type Spectinomycin Dihydrochloride
Chemical Name or Material Spectinomycin Dihydrochloride
Physical Form Powder/Solid
CAS 22189-32-8
Melting Point 210°C
Source Streptomyces spectabilis
Ignition Residue < 1.0%
Packaging Amber Glass
Specific Rotation 15 to 21°
Water 16.0 to 20.0%
Concentration >603μg/mg
Quantity 1 g
Molecular Formula C14H36Cl2N2O12
MDL Number MFCD00150886
Merck Index 15, 8866
InChI Key DCHJOVNPPSBWHK-UXXUFHFZSA-N
SMILES [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
IUPAC Name dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
Molecular Weight (g/mol) 495.34
PubChem CID 67108398
ChEBI CHEBI:9217
Formula Weight 495.3
Percent Purity >603 μg/mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.