Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Spectinomycin dihydrochloride pentahydrate | 22189-32-8 | 98.0% | 1 ML
SDP

Catalog No. 5000412153
Encompass_Preferred

Spectinomycin dihydrochloride pentahydrate is a broad-spectrum antibiotic that inhibits the growth of various gram-positive and gram-negative organisms. It functions by selectively targeting bacterial ribosomes and disrupting protein synthesis. Additionally, it acts as a noncompetitive inhibitor of td intron RNA with a Ki value of 7.2 mM.

  • Broad-spectrum antibiotic
  • Specific mechanism of action: selectively targets bacterial ribosomes to interrupt protein synthesis
  • Inhibits td intron RNA
  • Available in multiple forms
  • Research use only
  • High purity: guaranteed at 98.0% purity
  • Solubility: soluble in water at 62.5 mg/mL (126.17 mM)
  • Storage flexibility: can be stored at 4°C (sealed, away from moisture) for solid form; stock solutions can be stored at -80°C for 6 months or -20°C for 1 month
  • Detailed documentation

Catalog No. 50-004-12153 Supplier Medchemexpress LLC Supplier No. HYB1828A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spectinomycin dihydrochloride pentahydrate is a broad-spectrum antibiotic that inhibits the growth of various gram-positive and gram-negative organisms. It functions by selectively targeting bacterial ribosomes and disrupting protein synthesis. Additionally, it acts as a noncompetitive inhibitor of td intron RNA with a Ki value of 7.2 mM.

  • Broad-spectrum antibiotic
  • Specific mechanism of action: selectively targets bacterial ribosomes to interrupt protein synthesis
  • Inhibits td intron RNA
  • Available in multiple forms
  • Research use only
  • High purity: guaranteed at 98.0% purity
  • Solubility: soluble in water at 62.5 mg/mL (126.17 mM)
  • Storage flexibility: can be stored at 4°C (sealed, away from moisture) for solid form; stock solutions can be stored at -80°C for 6 months or -20°C for 1 month
  • Detailed documentation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.