Learn More
Alkali Scientific Spectinomycin Dihydrochloride Pentahydrate, 25 G
SDP
Catalog No. 5000660415
Encompass_Preferred
Spectinomycin Dihydrochloride Pentahydrate in a 25g powder form is a potent antibiotic used to combat bacterial infections, especially those caused by Gram-negative pathogens. It is primarily employed in clinical settings to treat Neisseria gonorrhoeae, particularly in penicillin-resistant cases. In addition to its therapeutic uses, spectinomycin is extensively utilized in laboratory research to inhibit protein synthesis in bacterial cultures. This allows for studies related to bacterial growth, gene expression, and antibiotic resistance. The 25g size is ideal for experimental setups requiring precise concentrations, providing sufficient material for both clinical and research applications.
Catalog No. 50-006-60415 Supplier Alkali Scientific Supplier No. SPE004CP
Spectinomycin Dihydrochloride Pentahydrate in a 25g powder form is a potent antibiotic used to combat bacterial infections, especially those caused by Gram-negative pathogens. It is primarily employed in clinical settings to treat Neisseria gonorrhoeae, particularly in penicillin-resistant cases. In addition to its therapeutic uses, spectinomycin is extensively utilized in laboratory research to inhibit protein synthesis in bacterial cultures. This allows for studies related to bacterial growth, gene expression, and antibiotic resistance. The 25g size is ideal for experimental setups requiring precise concentrations, providing sufficient material for both clinical and research applications.